Learn More
Alpha Packaging™ B-safe™ Polyethylene Biohazard Bag
Manufactured from polyethylene. Alpha Packaging™ B-safe™ Polyethylene Biohazard Bags effectively contain biohazard waste.
Brand:  Alpha Packaging™ 12300609
 View more versions of this product
Product Code. 12300609
  • £113.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from polyethylene. Alpha Packaging™ B-safe™ Polyethylene Biohazard Bags effectively contain biohazard waste.
  • Comes in loose packs or benchtop dispenser boxes.
  • For heavy-duty disposal
  • Marked on both sides with biohazard symbol and warnings:
    • "Biohazard, Danger of Infection"
    • "Burn Without Opening"
  • Warning in English, French, and German
  • Polyethylene (500 gauge)
Specifications
Specifications
Yellow
355 mm
Polyethylene
Single-use
255 mm
Biohazard Bag