Learn More
Alpha Packaging™ B-safe™ Polyethylene Biohazard Bag
Manufactured from polyethylene. Alpha Packaging™ B-safe™ Polyethylene Biohazard Bags effectively contain biohazard waste.
Brand: Alpha Packaging™ 12340609
Code : 04
Additional Details : Weight : 1.36000kg
Description
Manufactured from polyethylene. Alpha Packaging™ B-safe™ Polyethylene Biohazard Bags effectively contain biohazard waste.
- Comes in loose packs or benchtop dispenser boxes.
- For heavy-duty disposal
- Marked on both sides with biohazard symbol and warnings:
- "Biohazard, Danger of Infection"
- "Burn Without Opening"
- Warning in English, French, and German
- Polyethylene (500 gauge)
Spécification
Yellow | |
Yes | |
200 mm | |
Polyethylene |
200 x 200 x 250 mm | |
255 mm | |
200 mm | |
Biohazard Bag |