missing translation for 'onlineSavingsMsg'
Learn More

B-Tribromoborazine, 96%

Product Code. 11350328
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11350328

Brand: Thermo Scientific Alfa Aesar 043723.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13703-88-3 | B3Br3H3N3 | 317.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13703-88-3
Molecular Formula B3Br3H3N3
Molecular Weight (g/mol) 317.187
MDL Number MFCD00058924
InChI Key PCLMMJIWMGUYKF-UHFFFAOYSA-N
Synonym b-tribromoborazine, 2,4,6-tribromoborazine, b-tribromoborazol, borazine, 2,4,6-tribromo
PubChem CID 23278010
IUPAC Name 2,4,6-tribromo-1,3,5,2,4,6-triazatriborinane
SMILES B1(NB(NB(N1)Br)Br)Br

Specifications

Melting Point 124°C to 128°C
Color White
Physical Form Powder
Quantity 1 g
UN Number UN1759
Solubility Information Insoluble in water.
Formula Weight 317.2
Percent Purity 96%
Sensitivity Moisture sensitive
Chemical Name or Material B-Tribromoborazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.