Learn More
Motic™ BA210E Trinocular Compound Microscope
Use this microscope for both educational and teaching environments. Motic™ BA210E Trinocular Compound Microscope is ideal for a variety of life sciences and medical applications.
Brand:  Motic™ 1100100403761
Product Code. 17157401
  • £1220.00 / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Excellent spherical aberration correction to significantly improve field flatness and resolution.
  • Superior color fidelity through new multi-layer coated lenses.
  • Significantly increased working distances and lead-free manufacturing process according to RoHS standard.
  • The ELITE model's addition of a new rackless stage concept without prominent gear rack allows a more convenient use of the x/y movement.
  • Full freedom of Illumination options, the new model allows for easy interchangeability of its 6V/30W halogen bulb with LED modules
  • Motic Infinity Optics (CCIS™) allows a higher level of performance in education and training.
  • Contrast methods like phase contrast, polarization and dark field are easily performed using optional accessories.
Specifications
Specifications
100/240 V
CCIS EC Plan
Universal power supply 100/240 V (CE)
Trinocular compound microscope
High Eyepoint N-WF 10x/20 mm with diopter adjustment on both eyepieces
4X, 10X, 40XS, 100XS (oil)
Reversed quadruple revolving nosepiece.
Focusable Abbe condenser N.A. 1.25 with iris diaphragm and slot
Halogen with Intensity Control 6V/30w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Motic™ BA210E Trinocular Compound Microscope > Product Type: Trinocular Compound Microscope