missing translation for 'onlineSavingsMsg'
Learn More

Hartmann Bacillol™ Zero Tissues

Product Code. p-200110162
Click to view available options
Packaging Quantity:
Flow-Pack, 100 Tissues
Flow-Pack, 80 XL Tissues
Dimensions (L x W):
20 x 17 cm (7.9 x 6.7 in.)
20 x 30 cm (7.9 x 11.8 in.)
Size:
Standard
X-Large
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 30048900

Brand: Hartmann 981935

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Cleaning and disinfecting wipes based on organic acids, Bacillol™ Zero Tissues are highly effective and gentle on materials.

  • Ready-to-use quick disinfectant wipes made from 100% plastic-free material
  • Virucidal in 2 minutes
  • Free from alcohols, quaternary ammonium compounds, peroxides, perfumes, and dyes
  • >98.5% ingredients of organic origin
  • Patented formulation - organic acid complex
  • No hazardous substance labeling
  • Very good material compatibility
  • Sustainable, high-quality, non-woven wipe material
  • BPR-tested
  • Excellent, visible wetting of surfaces
  • Can be used without gloves
  • Good cleaning properties
  • Practical folding system for safe and easy removal of the individual wipes from the resealable flow-pack

Specifications

Product Type Disinfectant Wipe
Length (English) 7.9 in.
Length (Metric) 20 cm
Width (English) 6.7 in.
Width (Metric) 17 cm
Material Non-Woven Cloth
Autoclavable Not Autoclavable
Disposable Single-use
Dimensions (L x W) 20 x 17 cm (7.9 x 6.7 in.)
For Use With (Application) Cleaning and Disinfecting
Packaging Quantity Flow-Pack, 100 Tissues
Ply 1-ply
Size Standard
Sterility Sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.