Learn More
Novacyt - Microgen Bioproducts Bacillus ID
Microgen Bioprpducts Bacillus ID. For the identification of mesophilic Bacillus species isolated from food materials and the environment.
Brand:  Novacyt - Microgen Bioproducts MID-66
Product Code. 17323221
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Microgen Bioproducts Bacillus-ID has been developed for the identification of mesophilic Bacillus spp. and related genera isolated from food, food ingredients and beverages either as food spoilage or food poisoning organisms.
Specifications
Specifications
ISO13485 ISO9001
Yes
Arabinose, Cellobiose, Inositol, Mannitol, Mannose, Raffinose, Rhamnose, Salicin, Sorbitol, Sucrose, Trehalose, Xylose, Adonitol, Galactose, M-D-Mannopyranoside, M-D-Glucopyranoside, Inulin, Melezitose, Indole, Nitrate, Arginine, Citrate, Voges Proskauer
Bacterial Isolate
Test Strip
20 Test strips
Monera
20 x 3 mL bottles, 20 Test strips
0.98
Read after 24 hours and 48 hours incubation
2°C to 8°C
Non-waived
20 x 3 mL bottles, 20 Test strips
0.98
An Identification System for The Mesophilic Bacillus Species and Genera From Food and Clinical Sources
12 per test strip
Carton with pouches and strip holding tray
Mesophilic Bacillus species and Genera from Food and Clinical sources
Bacteria