missing translation for 'onlineSavingsMsg'
Learn More

IDEX Health & Science, LLC Back Pressure Regulator (BPR) Cartridge

Operates as an easy to use, replaceable component for BPR systems

Brand:  IDEX Health & Science, LLC 02017-26

Product Code. 13000440

  • £143.00 / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This replacement Back Pressure Regulator (BPR) Cartridge operates as an easy to use, replaceable component for BPR systems. This cartridge is constructed from PEEK, Tefzel (EFTE), PCTFE and includes a gold plated stainless steel spring that delivers consistent, reliable usage. Conveniently designed to be used with any standard BRP Holder and creates a back pressure of 750 psi (34 bar). This replacement cartridge has a black body and blue end cap.

  • Replacement Back Pressure Regulator (BPR) Cartridge
  • Operates as an easy to use component for BPR systems
  • Constructed from PEEK, Tefzel (EFTE), and PCTFE
  • Includes a gold plated stainless steel spring that delivers consistent, reliable usage
  • Conveniently designed to be used with any standard BRP Holder
  • Creates a back pressure of 750 psi (52 bar)
  • Swept Volume: 87 μL
Specifications

Specifications

Back Pressure Regulator Cartridge
750 psi (52 bar)
87 µL
Standard BRP Holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IDEX Health & Science, LLC Back Pressure Regulator (BPR) Cartridge > 750 psi (52 bar)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.