missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Bacteria protease native Protein from
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| For Use With (Application) | Enzyme Activity, SDS-PAGE |
| Name | Protease (Staphylococcus aureus) Native Protein |
| Preparation Method | Native protein isolated from the culture filtrate of Staphylococcus aureus, strain V8 |
| Quantity | 5 mg |
| Storage Requirements | Store at 4°C. This product is stable for at least 3 months. |
| Regulatory Status | RUO |
| Common Name | PROTEASE |
| Biological Activity | ≥ 500 units/mg. Unit definition: One unit will change A280nm 0.001 per minute at 37°C, pH 7.8, using casein as the substrate. |
| Species | Bacteria |
| Protein Tag | None |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction