missing translation for 'onlineSavingsMsg'
Learn More

Novacyt - Microgen Bioproducts Bacterial ID System Reagent/Consumable

Additional/supplemental reagent/consumable for biochemical identification system.

£27.40 - £56.30

Specifications

Product Type Biochemical Identification
For Use With (Application) Biochemical Identification Kits
View More Specs

Products 10
Product Code Brand ContentandStorage Price Quantity & Availability  
Product Code Brand ContentandStorage Price Quantity & Availability  
15967122
View Documents
Novacyt - Microgen Bioproducts
MID61G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15907122
View Documents
Novacyt - Microgen Bioproducts
MID61A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15917122
View Documents
Novacyt - Microgen Bioproducts
MID61B
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15927122
View Documents
Novacyt - Microgen Bioproducts
MID61C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15937122
View Documents
Novacyt - Microgen Bioproducts
MID61D
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15947122
View Documents
Novacyt - Microgen Bioproducts
MID61E
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15957122
View Documents
Novacyt - Microgen Bioproducts
MID61F
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15977122
View Documents
Novacyt - Microgen Bioproducts
MID61H
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15987122
View Documents
Novacyt - Microgen Bioproducts
MID61K
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15997122
View Documents
Novacyt - Microgen Bioproducts
MID61L
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Replacement/supplemental reagent/consumable.
  • From a range of simple, reliable, bacterial identification systems for food, environmental, and industrial laboratories.
  • These tests will arm professionals with the necessary information to maximize the effectiveness of appropriate patient management.
  • The range is expanding quickly and will encompass the widest range of microbiological testing solutions.
Specifications

Specifications

Biochemical Identification
Biochemical Identification Kits
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MID-61G

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : oxidase strips

SDS
Documents

Documents

Product Certifications