Learn More
Kartell™ L-Shaped Spreader
Kartell™ L-Shaped Spreader is great for dispersing liquids across agar culture plates and features a 100° angle for easier use.
Brand:  Kartell™ 0264800
Code : NEW
Additional Details : Weight : 3.40200kg
Product Code. 10768484
  • £431.00 / Pack of 400
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
  • Disposable
  • Sterile PS spreaders for bacteriology with special 100° angling for easier use
  • Especially handy for dispersing liquids across agar culture plates
Specifications
Specifications
L-Shape
5 pcs.
CE/IVD
White
Polystyrene