missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Bacto™ Brain Heart Infusion
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
Description
BHI broth is used for the cultivation of a wide variety of microorganisms, including bacteria (streptococci, pneumococci and meningococci), yeasts and molds. It serves as a base for supplemented media containing 0.1% agar, Fildes enrichment or 6.5% sodium chloride.
- To use dehydrated product: suspend 37g in 1L of purified water
- Displays a good cultural response to Neisseria meningitides 13090, Streptococcus pneumonia 6305 and Streptococcus pyogenes 19615
- pH (3.7% solution, 25°C): 7.4 ± 0.2
Certifications
AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA |
| Color | Light tan |
| Content And Storage | 2 to 25°C |
| Form | Dehydrated (free-flowing, homogenous) |
| Media Type | Brain Heart Infusion |
| Product Type | Brain Heart Infusion |
| Formulation | Calf brain (infusion from 200g) 7.7g/L, Beef heart (infusion from 250g) 9.8g/L, Proteose Peptone 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L |
| For Use With (Application) | Cultivation of fastidious and non-fastidious microorganisms, including aerobic and anaerobic bateria, yeasts and molds |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Difco™ Bacto™ Brain Heart Infusion >
Spot an opportunity for improvement?Share a Content Correction