Learn More
Thermo Scientific™ Bags and Liners for HyPerforma™ Single-Use Mixer DS 300, 200L Liner - Bottom drain, non-irradiated
The 200L open-top, non-irradiated BioProcess Container (BPC) tank liner with bottom drain is composed of CX3-9 single web, multi-layer plastic film and compatible with Thermo Scientific™ HyPerforma Single-Use Mixer DS 300.
Brand: Thermo Scientific™ SH30399.03
1415.44 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.45000kg
Description
The 200L open-top, non-irradiated BioProcess Container (BPC) tank liner with bottom drain is composed of CX3-9 single web, multi-layer plastic film and compatible with Thermo Scientific™ HyPerforma Single-Use Mixer DS 300. Implement industry-proven BPCs and liners in the overall process of shipping and storing your liquid-liquid and powder-liquid bioprocess mixing applications. BPCs and liners come in open- and closed-top design and are manufactured using our proprietary CX5-14 and CX3-9 film that easily fit in every plastic drum size.
Key Features of HyPerforma Single-Use Mixer DS 300 BPCs and liners:
• All standard BPCs and liners are designed to fit “out-of-the-box“
• Available in volume options of 50, 100, and 200L
• Applicable to open- and closed-top mixing applications
• Powder-liquid and liquid-liquid mixing
• World class CX5-14 and CX3-9 film
• Adaptable to multiple upstream process applications in media and buffer prep
Required Accessories:
• HyPerforma Single-Use Mixer DS 300 docking station
• HyPerforma SIngle-Use Mixer DS 300 drums
Recommended for:
• Powder-liquid or liquid-liquid mixing applications in single use
• High-throughput mixing and storage
• Additional configurations are available to suit your application requirements.
Specifications
200 L | |
CX3-9 | |
Packages placed in strong cardboard cartons with product and lot identification | |
Non-Irradiated | |
HyPerforma™ | |
Impeller Sleeve with Quick Connect: C-Flex™ Tubing |
Liner - Bottom drain, non-irradiated | |
Bottom Port: 0.5 in. | |
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area | |
Bottom Port: 1.27 x 1.91 x 122 cm (0.5 x 0.75 x 304.1 in.) Polycarbonate Quick Connect 12.7 mm (0.5 in.) insert (MPX) and cap | |
Fewer resources used and less waste |