missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Baird-Parker (ISO) Agar

Product Code. 15215862
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15215862

Brand: Thermo Scientific™ PO1195A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ensure accurate detection of Staphylococcus aureus with this agar, compliant with ISO standards for consistent and reliable microbial analysis.

Ensure accurate detection of Staphylococcus aureus with Baird-Parker (ISO) Agar, compliant with ISO standards for consistent and reliable microbial analysis.

TRUSTED_SUSTAINABILITY

Specifications

Color Opaque
Content And Storage 2°C to 12°C
Format 90 mm Plate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 17 g
Quantity 10 Plates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.