missing translation for 'onlineSavingsMsg'
Learn More
Learn More
J.T. Baker Baker-flex™, Cellulose DEAE, J.T. Baker™
Click to view available options
Quantity:
50 Sheets
Description
Flexible TLC Sheet
Specifications
Specifications
Color | White |
CAS | 9004-34-6 |
Physical Form | Crystal or Powder |
Solubility Information | Insoluble in water |
Quantity | 50 Sheets |
Odor | Odorless |
Chemical Name or Material | Cellulose DEAE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction