missing translation for 'onlineSavingsMsg'
Learn More
Learn More
J.T. Baker BAKERBOND spe™ Aromatic Sulfonic Acid (C6H5SO3H) Disposable Extraction Columns, J.T. Baker™
536.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
30/Box
50/Box
Description
Disposable Extraction Columns
Specifications
Odor | Odorless |
Quantity | 50/Box |
Physical Form | Solid |
Solubility Information | Practically Insoluble |
Chemical Name or Material | Aromatic Sulfonic Acid (C6H5SO3H) Disposable Extraction Columns |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'