Learn More
Avantor™ BAKERBOND Speedisk™ Oil and Grease Extraction Disk
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Oil and Grease Extraction Disk are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
Brand: Avantor™ 8060-06
536.75 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
- Require minimal glassware, set up time, and assembly
- Minimizes impact of extractable compounds
- Resists clogging and ensures high throughput rate
- Reduce SPE run times
Specifications
For Use with Slightly Polar to Non-Polar Hydrocarbons, EPA Method 1664, Rev. A. | |
Oil and Grease Extraction Disk | |
50 mm |
Expanded Extraction Station (8095-06) | |
20/Pk. |