missing translation for 'onlineSavingsMsg'
Learn More

Avantor™ BAKERBOND Speedisk™ SAX Quaternary Amine Extraction Disk

Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk SAX Quaternary Amine Extraction Disk are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.

Brand:  Avantor™ 8058-06

660.25 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.



Product Code. 15527694

  • £734.00 / Each
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Specifications

Specifications

EPA Method 552.1, Haloacetic Acids and Dalapon
SAX Quaternary Amine Extraction Disk
50 mm
Expanded Extraction Station (8095-06)
20/Pk.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diisopropyl ether

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Diisopropyl ether

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.