missing translation for 'onlineSavingsMsg'
Learn More

BICASA Balance Table (Vibration-proof)

Vibration-proof Balance Table with HPL countertop and marble plate.

Brand:  BICASA BT.1200.900.880

 View more versions of this product

Product Code. 30057592

  • £2205.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Specifically designed and manufactured to use with balances and analytical instruments
  • Marble vibration-proof central plate (Carrara White)
  • Fire-retardant particleboard filler body
  • HPL countertop
  • Light Grey color (2 decorative)
  • Separate internal welded tubular steel frame
  • with levelling feet and CAU devices
Specifications

Specifications

Balance Table
Not Autoclavable
Holding balances
Frame, Marble Plate (770 x 350 x 40 mm th)
1200 mm
Light Gray
900 x 1200 x 880 mm
Balances and analytical instruments
47.2 in.
Phenolic Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.