Learn More
Thermo Scientific Chemicals Ball pestle; clear plexiglas; 1/8in. diameter
For use with Wig-L-Bug Mixing/Grinding Mill (AA44204KT and AA44604KT)
Brand:  Thermo Scientific Chemicals 044532.KN
 View more versions of this product
Product Code. 11445280
  • £159.80 / Pack of 100
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100/Pk.
3.17mm
Wig-L-Bug Mixing/Grinding Mill (AA44204KT and AA44604KT)
0.125 in.
Ball Pestle