missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Balzers chrome layer etching solution, Fluka™
Balzers chrome layer etching solution
Brand: Honeywell Fluka 74727-200L
Code : NEW
Additional Details : Weight : 228.00000kg
Specifications
Balzers chrome layer etching solution | |
16774-21-3,64-19-7,7732-18-5 | |
Synthesis | |
Acidic |
Light Yellow | |
100°C (1.013 hPa) | |
Metal Drum | |
200 L |