Learn More
Uvex Bamboo TwinFlex™ D xg Cut Protection Gloves
Description
Bamboo TwinFlex™ D xg: Sustainability, comfort and sensitivity combined with high cut protection (CutD)
The patented Bamboo TwinFlex™ technology combines the best of both worlds in one fine 18-gauge cut protection glove (Cut Level D). On the inside of the glove, wearers experience noticeable comfort for their skin thanks to the bamboo-viscose. The smooth fit and high moisture absorption with a rapid action provides long-lasting comfort and high wearer acceptance. With a flexible fit thanks to the bamboo-viscose, the glove adapts precisely to fit the shape of the hand within 3 to 5 minutes of wearing. High-performance fibers offer a high level of protection against abrasion and cut hazards. The additional protection zone in the area between the thumb and index finger provides double the abrasion resistance (200%) compared to the inside of the glove to increase its service life/durability. Thanks to the Xtra-Grip aqua-polymer foam coating, the Bamboo TwinFlex™ D xg offers excellent grip in dry and slightly damp/oily conditions. In addition, the Bamboo TwinFlex D xg features a total of 45% sustainable materials: recycled polyamide and bamboo-viscose, which is based on renewable raw materials.
General Features:
- Excellent sensitivity in cut protection level D
- High level of wearer comfort thanks to bamboo fibre
- 45% sustainable materials: recycled polyamide and bamboo viscose, which is based on renewable raw materials
- High efficiency due to long service life, all thanks to protection + premium Xtra-Grip coating
- CO2-neutral production in Germany, free from solvents
Protective Features
- Certified to EN 388:2016 (4 X 4 1 D) and to EN 407 (X 1 X X X X)
- Dermatologically tested for skin compatibility, free from allergenic accelerators
- Particularly light, thin, and flexible with high cut protection (4X41D according to EN 388:2016)
- No need to remove the glove thanks to touchscreen capability protection zone with smooth material surface ensures trouble-free workflow in accordance with EN 407, the model is suitable for contact warmth up to +100 °C
Comfort Features
- Patented Bamboo TwinFlex™ yarn technology
- Silky soft feel against the skin
- Fast absorption and high retention of moisture
- Adaptive fit: Adapts precisely to fit the shape of any hand within 3 to 5 minutes of wearing
Areas of application
- Installation of sharp-edged or unburred parts
- Glass and metalworking industry, automotive industry
- Precision work
- Quality control
- Areas with use of touchscreens food processing
Specifications
Specifications
Certifications/Compliance (Europe) | EN 388:2016 (4 X 4 1 D), EN 407 (X 1 X X X X) |
Construction | Seamless Knit |
Coating Material | Aqua-Polymer |
Color | Green |
Hand | Ambidextrous |
Latex Free | Yes |
Material | Bamboo-viscose, Elastane, HPPE, Polyamide, Steel |
Product Type | Cut-resistant Gloves |
Size | 7 |
EN388 Standard | EN 388:4 X 4 1 D |
Show More |