missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BamHI (10 U/μL)

The BamHI restriction enzyme recognizes G^GATCC sites and cuts best at 37°C in its own unique buffer.

Brand:  Thermo Scientific™ ER0052

88.35 GBP valid until 2024-12-31
Use promo code "21978" to get your promotional price.


 View more versions of this product

Product Code. 11843823

  • £93.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality – stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities

Applications

  • Molecular cloning
  • Restriction site mapping
  • Genotyping
  • Southern Blot
  • Restriction fragment length polymorphism (RFLP)
  • SNP
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 U/μL
BamH I
Yes
No
Restriction Enzyme
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Unique Buffer (10X Buffer BamHI)
37°C
5 x 4000 U
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.