missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAPTA, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Selective calcium chelator.
Specifications
CAS | 85233-19-8 |
Molecular Formula | C22H24N2O10 |
Quantity | 100 mg |
Synonym | bapta, bapeta, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, ca ii-bapta, 1,2-bis-2-aminophenoxy ethane n,n,n',n'-tetraacetic acid, unii-k22ddw77c0, 1,2-bis o-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, bapta tetrasodium salt hydrate, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-carboxymethyl, 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetic acid |
InChI Key | FTEDXVNDVHYDQW-UHFFFAOYSA-N |
SMILES | C1=CC=C(C(=C1)N(CC(=O)O)CC(=O)O)OCCOC2=CC=CC=C2N(CC(=O)O)CC(=O)O |
IUPAC Name | 2-[2-[2-[2-[bis(carboxymethyl)amino]phenoxy]ethoxy]-N-(carboxymethyl)anilino]acetic acid |
Molecular Weight (g/mol) | 476.438 |
PubChem CID | 104751 |
ChEBI | CHEBI:60888 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'