missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barbituric Acid, BAKER™, J.T. Baker™
179.55 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Molecular Formula | C4H4N2O3 |
MDL Number | MFCD00006666 |
Synonym | barbituric acid, malonylurea, 2,4,6 1h,3h,5h-pyrimidinetrione, 6-hydroxyuracil, pyrimidinetrione, 2,4,6-trihydroxypyrimidine, 2,4,6-pyrimidinetrione, pyrimidine-2,4,6 1h,3h,5h-trione, barbiturate, malonylharnstoff |
InChI Key | HNYOPLTXPVRDBG-UHFFFAOYSA-N |
SMILES | O=C1CC(=O)NC(=O)N1 |
IUPAC Name | 1,3-diazinane-2,4,6-trione |
Molecular Weight (g/mol) | 128.09 |
Quantity | 100 g |
Packaging | Wide Mouth Amber Glass Bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'