Learn More
Corning™ Bar-coded Cryogenic Vials
For storing biological and compound samples in labeled vials at cryogenic temperatures. Corning™ Bar-coded Cryogenic Vials provides a validated unique bar-code that will not fade or fall off.
£345.58 - £541.96
Especificaciones
Volume (Metric) | 2 mL |
---|---|
For Use With (Application) | Cell biology, cell banking, compound cryopreservation |
Código de producto | Marca | Barcode | Thread Style | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Barcode | Thread Style | Precio | Cantidad y disponibilidad | |||||
15595719
|
Corning™
8670 |
2D | Internal |
£535.00
Pack of 500 |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||||
15505729
|
Corning™
8671 |
2D | External threaded |
£541.96
Pack of 500 |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||||
15515729
|
Corning™
8672 |
1D | Internal |
£351.00
Pack of 500 |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||||
15545729
|
Corning™
8676 |
1D | External |
£345.58
Pack of 500 |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||||
Descripción
- Manufactured from polypropylene
- Leak proof seal for integrity of storage
- Autoclavable vials and caps, can be re-sterilized
- Unique 1D, 2D bar-codes and human readable characters
- Features star shaped cavity in cap and is compatible with decapping equipment
- Locking base stabilizes tubes for automated handling
- Graduations and writing patch for manual use also
- Each tube is checked for clarity and readability of 2D code
Especificaciones
2 mL | |
Cell biology, cell banking, compound cryopreservation |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Corning™ Bar-coded Cryogenic Vials