Learn More
Thermo Scientific™ Barcoded Latch Racks for Matrix™ Storage Tubes
Create custom arrays with empty ANSI-format racks featuring 2D and linear barcodes and human readable type on three sides and a 2D barcode on the base of the rack.
£5.30 - £237.00
Product Code | Brand | For Use With (Equipment) | No. per Case | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | No. per Case | Price | Quantity & Availability | ||||
15543734
|
Thermo Scientific™
4897-BR |
1.0mL Screwtop Tubes | 5 |
£121.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
15553734
|
Thermo Scientific™
4898-BR |
500μL Tubes | 10 |
£237.00
Pack of 10 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
15563734
|
Thermo Scientific™
4900-BR |
500μL Screwtop Tubes | 5 |
£121.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
11620833
|
Thermo Scientific™
4902 |
1.4mL tubes | 10 |
£63.35
Pack of 10 |
In Stock
Log in to see stock. |
||||
15374297
|
Thermo Scientific™
4903 |
0.75mL tubes | 10 |
£5.30
Each |
In Stock
Log in to see stock. |
||||
Description
Enhance sample storage at low temperatures, including vapor phase Liquid Nitrogen, with Thermo Scientific™ Matrix™ barcoded latch racks. Barcoded Latch Racks feature a linear barcode, 2D barcode and human readable rack identification on three sides of the rack for the ultimate in sample tracking. Use in combination with Thermo Scientific 2D barcoded tubes, barcode readers, arrayers and capping equipment.
Superior Storage Rack Design
- Storage in robot-friendly ANSI-footprint rack
- Racks feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking regardless of your barcode system
- Rack barcodes are compatible with our range of VisionMate readers
- Hinged lid rests in “open” position on rack while conducting manual pipetting to prevent contamination of lid sitting on bench
Latch Racks Save Precious Space
- ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
- Latch rack lid can be lifted robotically for tube access with high-throughput automated applications