missing translation for 'onlineSavingsMsg'
Learn More

Barium 2-ethylhexanoate, 20% w/w in 2-ethylhexanoic acid, Thermo Scientific™

Product Code. 11365098
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2457-01-4
MDL Number MFCD00058696

Specifications

Odor Weak, characteristic
Molecular Formula C16H30BaO4
Quantity 50 g
Synonym barium 2-ethylhexanoate, barium bis 2-ethylhexanoate, barium 2+ ; 2-ethylhexanoate, hexanoic acid, 2-ethyl-, barium salt 2:1, acmc-20alrk, hexanoic acid, 2-ethyl-, barium salt, dsstox_cid_24888, dsstox_rid_80561, dsstox_gsid_44888, bis 2-ethylhexanoic acid barium salt
InChI Key VJFFDDQGMMQGTQ-UHFFFAOYNA-L
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ba+2]
IUPAC Name barium(2+);2-ethylhexanoate
Molecular Weight (g/mol) 423.74
PubChem CID 102821
Formula Weight 423.76
Concentration or Composition (by Analyte or Components) 20% w/w in 2-ethylhexanoic acid
Physical Form Liquid
Chemical Name or Material Barium 2-ethylhexanoate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'