missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium acetate, 99+%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
pH | 7 to 8.5 (at 5% 20°C) |
Physical Form | Crystalline Powder |
Quantity | 500 g |
Assay Percent Range | 99.0 to 102% (Complexometry) |
Linear Formula | (CH3CO2)2Ba |
Merck Index | 15, 961 |
Solubility Information | Solubility in water: soluble. Other solubilities: practically insoluble in alcohol |
Formula Weight | 255.43 |
Percent Purity | ≥99% |
Show More |
Safety and Handling
Product Identifier
- Barium acetate
- Warning
- Acute toxicity Category 4
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'