missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, Puriss. p.a., ACS Reagent, ≥99%, Honeywell Fluka™

Product Code. 15603800
Click to view available options
Quantity:
250 g
1 kg
50 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

Quantity 250 g
Linear Formula (CH3COO)2Ba
UN Number NONH for all modes of transport
Beilstein 3693411
IUPAC Name barium(2+);diacetate
Formula Weight 255.42g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Barium Acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'