Learn More
Barium acetate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
£41.71 - £488.43
Chemical Identifiers
CAS | 543-80-6 |
---|---|
Molecular Formula | C4H6BaO4 |
Molecular Weight (g/mol) | 255.42 |
MDL Number | MFCD00012447 |
InChI Key | ITHZDDVSAWDQPZ-UHFFFAOYSA-L |
Synonym | barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetateShow More |
PubChem CID | 10980 |
SMILES | [Ba++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366148
|
Thermo Scientific Alfa Aesar
011126.06 |
5 g |
£41.71
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376148
|
Thermo Scientific Alfa Aesar
011126.14 |
25 g |
£146.73
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356148
|
Thermo Scientific Alfa Aesar
011126.22 |
100 g |
£488.43
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
543-80-6 | |
255.42 | |
ITHZDDVSAWDQPZ-UHFFFAOYSA-L | |
10980 |
C4H6BaO4 | |
MFCD00012447 | |
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetateShow More | |
[Ba++].CC([O-])=O.CC([O-])=O |
Specifications
543-80-6 | |
Crystalline | |
Ba(OOCCH3)2 | |
UN1564 | |
14,965 | |
ITHZDDVSAWDQPZ-UHFFFAOYSA-L | |
barium(2+) diacetate | |
10980 | |
99.999% | |
Odorless | |
(metals basis) |
White | |
C4H6BaO4 | |
MFCD00012447 | |
3693411 | |
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate | |
[Ba++].CC([O-])=O.CC([O-])=O | |
255.42 | |
255.43 | |
Puratronic™ | |
2.47g/mL | |
Barium acetate |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'rtecsNumber' : AF4550000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.