missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide, anhydrous, 99%, Thermo Scientific Chemicals

£66.23 - £283.41

Chemical Identifiers

CAS 10553-31-8
MDL Number MFCD00003444
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307218
View Documents
Thermo Scientific Alfa Aesar
016792.18
50 g
£66.23
50g
Please to purchase this item. Need a web account? Register with us today!
11397208
View Documents
Thermo Scientific Alfa Aesar
016792.30
250 g
£283.41
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium bromide, anhydrous is used as the chemical intermediate, and the spice cosmetics chemical additive. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10553-31-8
MFCD00003444

Specifications

850°C
White
MFCD00003444
14,968
297.14
Odorless
4.781 g/mL
10553-31-8
Powder
UN1564
Soluble in water
99%
Hygroscopic
Barium bromide, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

missing translation for 'einecsNumber' : 234-140-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only