Learn More
Barium bromide dihydrate, 98+%, Thermo Scientific Chemicals
£32.80 - £103.00
Chemical Identifiers
CAS | 7791-28-8 |
---|---|
Molecular Formula | BaBr2H4O2 |
Molecular Weight (g/mol) | 333.17 |
MDL Number | MFCD00149153 |
InChI Key | HQQMRSGBIPFNSN-UHFFFAOYSA-L |
IUPAC Name | barium(2+) dihydrate dibromide |
SMILES | O.O.[Br-].[Br-].[Ba++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15463137
|
Thermo Scientific Alfa Aesar
A16328.22 |
100 g |
£32.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15473137
|
Thermo Scientific Alfa Aesar
A16328.36 |
500 g |
£103.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7791-28-8 | |
333.17 | |
HQQMRSGBIPFNSN-UHFFFAOYSA-L | |
O.O.[Br-].[Br-].[Ba++] |
BaBr2H4O2 | |
MFCD00149153 | |
barium(2+) dihydrate dibromide |
Specifications
7791-28-8 | |
MFCD00149153 | |
14,968 | |
O.O.[Br-].[Br-].[Ba++] | |
333.17 | |
≥98% |
BaBr2H4O2 | |
UN1564 | |
HQQMRSGBIPFNSN-UHFFFAOYSA-L | |
barium(2+) dihydrate dibromide | |
333.17 (297.15 Anhydrous) | |
Barium bromide dihydrate |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only