missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Barium bromide is a precursor to chemicals used in photography and to other bromides. Barium bromide was used to purify radium in a process of fractional crystallization devised by Marie Curie. Since radium precipitates preferentially in a solution of barium bromide, the ratio of radium to barium in the precipitate would be higher than the ratio in the solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Melting Point | 850°C |
Color | White |
Physical Form | Beads |
Quantity | 5 g |
UN Number | UN1564 |
Merck Index | 14,968 |
Solubility Information | It is soluble in water. |
Formula Weight | 297.14 |
Percent Purity | 99.998% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'