missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium calcium copper oxide (2-2-3), 99.9% (metals basis), Thermo Scientific™
£20.90 - £59.17
Specifications
Chemical Name or Material | Barium calcium copper oxide (2-2-3) |
---|---|
Physical Form | Powder |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD00211652 |
UN Number | UN1564 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Barium calcium copper oxide (2-2-3) | |
(metals basis) | |
UN1564 |
Powder | |
MFCD00211652 | |
99.9% |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures