missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, 99.95% (metals basis), Thermo Scientific Chemicals

£37.59 - £711.54

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342408
View Documents
Thermo Scientific Alfa Aesar
014452.09
10 g
£37.59
10g
Please to purchase this item. Need a web account? Register with us today!
11322408
View Documents
Thermo Scientific Alfa Aesar
014452.18
50 g
£64.32
50g
Please to purchase this item. Need a web account? Register with us today!
11352408
View Documents
Thermo Scientific Alfa Aesar
014452.30
250 g
£238.19
250g
Please to purchase this item. Need a web account? Register with us today!
11332408
View Documents
Thermo Scientific Alfa Aesar
014452.A1
1 kg
£711.54
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium carbonate is frequently employed in ceramic industry as a component of glazes. It is added to clay to precipitate some soluble salts while manufacturing bricks, tiles and potteries. Barium carbonate is mainly used as a raw material in the electrical engineering industries for glass production, and as a raw material in the magnet manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

513-77-9
197.34
AYJRCSIUFZENHW-UHFFFAOYSA-L
10563
CBaO3
MFCD00003448
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
[Ba++].[O-]C([O-])=O

Specifications

(decomposition)
513-77-9
Lump/Powder
CBaO3
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium(2+) carbonate
10563
Odorless
4.43
811°C
White
99.95%
MFCD00003448
Insoluble in water. Soluble in most of the acids,with the exception of sulfuric acid.
[Ba++].[O-]C([O-])=O
197.34
197.35
Off-white lump/powder
Barium carbonate
Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 208-167-3

missing translation for 'rtecsNumber' : CQ800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only