Learn More
Barium carbonate, 99+%, for analysis, Thermo Scientific Chemicals
£51.10 - £158.00
Chemical Identifiers
CAS | 513-77-9 |
---|---|
Molecular Formula | CBaO3 |
Molecular Weight (g/mol) | 197.34 |
MDL Number | MFCD00003448 |
InChI Key | AYJRCSIUFZENHW-UHFFFAOYSA-L |
Synonym | barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p |
PubChem CID | 10563 |
SMILES | [Ba++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10412221
|
Thermo Scientific Acros
217495000 |
500 g | Plastic bottle |
£51.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10217363
|
Thermo Scientific Acros
217490025 |
2.5 kg | Plastic bottle |
£158.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
513-77-9 | |
197.34 | |
AYJRCSIUFZENHW-UHFFFAOYSA-L | |
10563 |
CBaO3 | |
MFCD00003448 | |
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p | |
[Ba++].[O-]C([O-])=O |
Specifications
1450.0°C | |
513-77-9 | |
Powder | |
CBaO3 | |
MFCD00003448 | |
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p | |
AYJRCSIUFZENHW-UHFFFAOYSA-L | |
barium(2+) carbonate | |
10563 | |
≥99% | |
4.43 | |
Barium carbonate, p.a. |
811.0°C | |
White | |
99% min. (Complexometry) | |
BaCO3 | |
15, 965 | |
Solubility in water: 0.002g/100mL (20°C) | |
[Ba++].[O-]C([O-])=O | |
197.34 | |
197.35 | |
Analytical | |
4.4300g/mL |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 208-167-3
missing translation for 'rtecsNumber' : CQ8600000
missing translation for 'tsca' : TSCA
RUO – Research Use Only