Learn More
Barium carbonate, tech., Thermo Scientific Chemicals
Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers
£20.70 - £252.26
Chemical Identifiers
CAS | 513-77-9 |
---|---|
Molecular Formula | CBaO3 |
Molecular Weight (g/mol) | 197.34 |
MDL Number | MFCD00003448 |
InChI Key | AYJRCSIUFZENHW-UHFFFAOYSA-L |
Synonym | barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p |
PubChem CID | 10563 |
SMILES | [Ba++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11355797
|
Thermo Scientific Alfa Aesar
043478.22 |
100 g |
£20.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365797
|
Thermo Scientific Alfa Aesar
043478.36 |
500 g |
£47.64
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11385797
|
Thermo Scientific Alfa Aesar
043478.A3 |
2 kg |
£79.90
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11375797
|
Thermo Scientific Alfa Aesar
043478.A9 |
10 kg |
£252.26
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers.Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
513-77-9 | |
197.34 | |
AYJRCSIUFZENHW-UHFFFAOYSA-L | |
10563 |
CBaO3 | |
MFCD00003448 | |
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p | |
[Ba++].[O-]C([O-])=O |
Specifications
(decomposition) | |
513-77-9 | |
CBaO3 | |
14,969 | |
Almost insoluble in water; Soluble in NH4Cl solution,dilute HCl,HNO3,acetic acid; Insoluble in most alkalies;Soluble in most acids,with the exception of sulfuric acid; It is insoluble in water and ethanol. | |
[Ba++].[O-]C([O-])=O | |
197.34 | |
197.35 | |
Odorless | |
Barium carbonate |
811°C | |
Powder | |
MFCD00003448 | |
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p | |
AYJRCSIUFZENHW-UHFFFAOYSA-L | |
barium(2+) carbonate | |
10563 | |
Technical | |
4.43 |
Safety and Handling
GHS H Statement
H302;
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 208-167-3
missing translation for 'rtecsNumber' : CQ8600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only