missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride, 0.5N Standardized Solution, Thermo Scientific Chemicals

Product Code. 11333687
Click to view available options
Quantity:
1 L
4 L

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-37-2
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003445
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
Synonym barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
PubChem CID 25204
ChEBI CHEBI:63317
IUPAC Name barium(2+);dichloride
SMILES [Cl-].[Cl-].[Ba++]

Specifications

Color Colorless
Concentration 0.5N
Formula Weight 208.25
Odor Odorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Barium chloride, Standardized Solution
compliance-icons
Product Identifier
  • Barium chloride, 0.5N Standardized Solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Barium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.