missing translation for 'onlineSavingsMsg'
Learn More
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals Product Code.: 11469020

Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals

Product Code. 11469020
1000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11469020

Brand: Thermo Scientific Alfa Aesar A12905.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

Solubility
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.

Notes
Stable and hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7dShow MoreShow Less
PubChem CID 5284346
ChEBI CHEBI:86153
SMILES O.O.[Cl-].[Cl-].[Ba++]

Especificaciones

Melting Point 113°C (in 2H2O)
Quantity 1000 g
UN Number UN1564
Merck Index 14,971
Solubility Information Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.
IUPAC Name barium(2+);dichloride;dihydrate
Formula Weight 244.28 (208.25 Anhydrous)
Percent Purity ≥99%
Odor Odorless
Density 3.097
Chemical Name or Material Barium chloride dihydrate
Mostrar más Mostrar menos

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW09K015 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog NumberA12905.0B,A12905.0I,A12905.30
Lot NumberS14K032 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog NumberA12905.0B,A12905.0I,A12905.30
Lot NumberR21K020 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA12905.0B,A12905.0I,A12905.30
Lot Number10249210 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA12905.0B,A12905.0I,A12905.30
Lot NumberP29K026 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog NumberA12905.0B,A12905.0I,A12905.30
compliance-icons
Nombre del producto
  • Barium chloride dihydrate
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Indicaciones de peligro
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : Barium chloride (BaCl2), dihydrate

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado