missing translation for 'onlineSavingsMsg'
Learn More
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals Product Code.: 11489020

Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals

Product Code. 11489020
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11489020

Brand: Thermo Scientific Alfa Aesar A12905.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

Solubility
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.

Notes
Stable and hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7dShow MoreShow Less
PubChem CID 5284346
ChEBI CHEBI:86153
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Melting Point 113°C (in 2H2O)
Quantity 250 g
UN Number UN1564
Merck Index 14,971
Solubility Information Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.
IUPAC Name barium(2+) dihydrate dichloride
Formula Weight 244.28 (208.25 Anhydrous)
Percent Purity ≥99%
Odor Odorless
Density 3.097
Chemical Name or Material Barium chloride dihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW09K015 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog NumberA12905.0B,A12905.0I,A12905.30
Lot NumberS14K032 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog NumberA12905.0B,A12905.0I,A12905.30
Lot NumberR21K020 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA12905.0B,A12905.0I,A12905.30
Lot Number10249210 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA12905.0B,A12905.0I,A12905.30
Lot NumberP29K026 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog NumberA12905.0B,A12905.0I,A12905.30
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    3

    Hours

    19

    Minutes

    58

    Seconds

    51

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.