missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride Dihydrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Description
Barium Chloride Dihydrate, Certified AR for Analysis
Specifications
Specifications
Boiling Point | 1560°C |
Melting Point | 962°C |
Color | White |
pH | 5 |
Physical Form | Solid |
Quantity | 5 kg |
Formula Weight | 244.28g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Barium chloride dihydrate
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'