missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride Dihydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 12696667 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Unit Size:
1kg
3kg
500g
5kg
This item is not returnable. View return policy

Product Code. 12696667

Brand: Fisher Chemical B/0550/63

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Barium Chloride Dihydrate, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Boiling Point 1560°C
Melting Point 962°C
Color White
pH 5
Physical Form Solid
Quantity 3 kg
Formula Weight 244.28g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Barium chloride dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.