missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium chloride dihydrate, Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
4 x 5 kg
50 kg
6 x 1 kg
Beschreibung
- Exceeds standard ACS specifications with additional trace metals analysis
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Spezifikation
Spezifikation
Boiling Point | 1,560°C |
CAS Max % | 100.0000% |
Quantity | 50 kg |
Linear Formula | BaCl2 · 2H2O |
UN Number | UN1564 |
Formula Weight | 244.26 |
Percent Purity | >99% |
Grade | ACS Reagent/Reagent ISO/Reagent Ph. Eur. |
Packaging | Fiberboard Box |
Density | 3.100 g/cm3 |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Barium chloride dihydrate
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'