missing translation for 'onlineSavingsMsg'
Learn More
Barium chloride dihydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals Product Code.: 11330849

Barium chloride dihydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Product Code. 11330849
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11330849

Brand: Thermo Scientific Alfa Aesar 010646.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Melting Point 113°C-2H2O
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.
Formula Weight 244.28 (208.25 Anhydrous)
Odor Odorless
Appearance White powder
Density 3.097
Chemical Name or Material Barium chloride dihydrate, dihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium chloride dihydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.