missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride EU Pharmacopoeia, Fisher Chemical™

Catalog Number 12980594
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 101-27-9
Quantity 1 mL
Molecular Formula C11H9Cl2NO2
Synonym barban, barbamate, barbane, carbine, carbyne, chlorinat, carbin, karbin, neoban, carbyne herbicide
InChI Key MCOQHIWZJUDQIC-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Cl)NC(=O)OCC#CCCl
IUPAC Name 4-chlorobut-2-ynyl N-(3-chlorophenyl)carbamate
Molecular Weight (g/mol) 258.098
PubChem CID 7551
ChEBI CHEBI:33421
Concentration or Composition (by Analyte or Components) 100μg/mL
Packaging Ampoule
Chemical Name or Material Barban
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'