Learn More
Barium chromate, 98%, Thermo Scientific Chemicals
£49.25 - £142.71
Chemical Identifiers
CAS | 10294-40-3 |
---|---|
Molecular Formula | BaCrO4 |
Molecular Weight (g/mol) | 253.32 |
MDL Number | MFCD00014180 |
InChI Key | QFFVPLLCYGOFPU-UHFFFAOYSA-N |
Synonym | barium chromate, lemon chrome, permanent yellow, barium chromate vi, baryta yellow, ultramarine yellow, pigment yellow 31, ci pigment ylllow 31, barium chromate 1:1, unii-7d7o9cf0ix |
PubChem CID | 25136 |
SMILES | [Ba++].[O-][Cr]([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356978
|
Thermo Scientific Alfa Aesar
014669.30 |
250 g |
£49.25
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346978
|
Thermo Scientific Alfa Aesar
014669.A1 |
1 kg |
£142.71
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium chromate is used as a pigment, oxidizing agent, colorant in glass, and sulfate scavenger in chromium electroplating baths, ceramics, and porcelain. It acts as a corrosion inhibitor in jointing pastes and metal primers. Further, it is used in fuses, safety matches, ignition control devices and high-temperature batteries. In addition to this, it is used as a carrier for the chromium ions and useful as a burn rate modifier in pyrotechnic compositions. It is involved as a catalyst in the alkane dehydration reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-40-3 | |
253.32 | |
QFFVPLLCYGOFPU-UHFFFAOYSA-N | |
25136 |
BaCrO4 | |
MFCD00014180 | |
barium chromate, lemon chrome, permanent yellow, barium chromate vi, baryta yellow, ultramarine yellow, pigment yellow 31, ci pigment ylllow 31, barium chromate 1:1, unii-7d7o9cf0ix | |
[Ba++].[O-][Cr]([O-])(=O)=O |
Specifications
10294-40-3 | |
Powder | |
BaCrO4 | |
barium chromate, lemon chrome, permanent yellow, barium chromate vi, baryta yellow, ultramarine yellow, pigment yellow 31, ci pigment ylllow 31, barium chromate 1:1, unii-7d7o9cf0ix | |
QFFVPLLCYGOFPU-UHFFFAOYSA-N | |
barium(2+);dioxido(dioxo)chromium | |
25136 | |
Odorless | |
Barium chromate |
Yellow | |
98% | |
MFCD00014180 | |
Insoluble in water,acetic acid and chromic acid. | |
[Ba++].[O-][Cr]([O-])(=O)=O | |
253.32 | |
253.33 | |
4.5 g/cm3 |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P210-P220-P221-P261-P264b-P270-P271-P281-P285-P301+P312-P304+P340-P308+P313-P330-P342+P311-P370+P378q-P501c
H272-H302+H332-H334-H350
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III
missing translation for 'einecsNumber' : 233-660-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only