missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium hexafluoro-2,4-pentanedionate, Thermo Scientific™
£31.05 - £113.57
Chemical Identifiers
CAS | 118131-57-0 |
---|---|
Molecular Formula | C10H2BaF12O4 |
Molecular Weight (g/mol) | 551.43 |
MDL Number | MFCD00075633 MFCD00075633 |
InChI Key | WUXISMDFUAVWSS-PAMPIZDHSA-L |
Synonym | barium 2+ 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate, barium 2+ ion 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate |
PubChem CID | 44717251 |
IUPAC Name | barium(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate) |
SMILES | [Ba++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322458
|
Thermo Scientific Alfa Aesar
014503.03 |
1 g |
£31.05
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312458
|
Thermo Scientific Alfa Aesar
014503.06 |
5 g |
£113.57
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
118131-57-0 | |
551.43 | |
WUXISMDFUAVWSS-PAMPIZDHSA-L | |
44717251 | |
[Ba++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F |
C10H2BaF12O4 | |
MFCD00075633 MFCD00075633 | |
barium 2+ 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate, barium 2+ ion 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate | |
barium(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate) |
Specifications
220°C (decomposition) | |
Powder | |
MFCD00075633 MFCD00075633 | |
barium 2+ 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate, barium 2+ ion 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate | |
[Ba++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F | |
551.43 | |
551.44 |
118131-57-0 | |
C10H2BaF12O4 | |
UN1564 | |
WUXISMDFUAVWSS-PAMPIZDHSA-L | |
barium(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate) | |
44717251 | |
Barium hexafluoro-2,4-pentanedionate |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures