missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide, anhydrous, 94-98%

Product Code. 11306598
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11306598

Brand: Thermo Scientific Alfa Aesar 012195.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17194-00-2 | BaH2O2 | 171.341 g/mol

Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17194-00-2
Molecular Formula BaH2O2
Molecular Weight (g/mol) 171.341
MDL Number MFCD00003443
InChI Key RQPZNWPYLFFXCP-UHFFFAOYSA-L
Synonym barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous
PubChem CID 6093286
ChEBI CHEBI:32592
IUPAC Name barium(2+);dihydroxide
SMILES [OH-].[OH-].[Ba+2]

Specifications

Melting Point 408°C
Physical Form Powder
Quantity 250 g
Assay Percent Range 94 to 98%
Solubility Information Soluble in water.
Formula Weight 171.36
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material Barium hydroxide, anhydrous, anhydrous
compliance-icons compliance-icons
Product Identifier
  • Barium hydroxide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.