Learn More
Barium hydroxide octahydrate, 97%, Thermo Scientific Chemicals
Alfa Aesar Barium hydroxide octahydrate, 97%, Quantity: 500g, CAS: 12230-71-6, Melting Point: 78 deg.C, Molecular Formula: BaH18O10, Molecular Weight: 315.461, Percent Purity: 97%
£23.72 - £252.48
Chemical Identifiers
CAS | 12230-71-6 |
---|---|
Molecular Formula | BaH18O10 |
Molecular Weight (g/mol) | 315.46 |
MDL Number | MFCD00149152 |
InChI Key | ZUDYPQRUOYEARG-UHFFFAOYSA-L |
Synonym | barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrateShow More |
PubChem CID | 17749109 |
SMILES | O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430643
|
Thermo Scientific Alfa Aesar
A12714.36 |
500 g |
£23.72
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440643
|
Thermo Scientific Alfa Aesar
A12714.0E |
2500 g |
£81.53
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450643
|
Thermo Scientific Alfa Aesar
A12714.0C |
10,000 g |
£252.48
10000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium hydroxide octahydrate has been used in the centromeric heterochromatin banding technnique and to synthesize BaTiO3. The form Barium hydroxide was reported to catalyze the rate of β elimination of phosphoserine residues. It is also used as a general purpose additive for lubricants and greases, sugar fabrication, manufacturing soaps, fat saponification and chemical synthesis of other barium compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBarium hydroxide octahydrate has been used in the centromeric heterochromatin banding technnique and to synthesize BaTiO3. The form Barium hydroxide was reported to catalyze the rate of β elimination of phosphoserine residues. It is also used as a general purpose additive for lubricants and greases, sugar fabrication, manufacturing soaps, fat saponification and chemical synthesis of other barium compounds.
Solubility
Soluble in water (72 mg/ml at 20°C), methanol, ethanol (slightly), and water (56 mg/ml at 15°C). Insoluble in acetone (practically)
Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
Chemical Identifiers
12230-71-6 | |
315.46 | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
17749109 |
BaH18O10 | |
MFCD00149152 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrateShow More | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
Specifications
78°C | |
BaH18O10 | |
MFCD00149152 | |
14,977 | |
Soluble in water (72mg/ml at 20°C),methanol,ethanol (slightly),and water (56mg/ml at 15°C). Insoluble in acetone (practically) | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] | |
315.46 | |
315.48 (171.36 Anhydrous) | |
Odorless | |
2.18 |
12230-71-6 | |
Ba(OH)2·8H2O | |
UN3262 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
barium(2+);dihydroxide;octahydrate | |
17749109 | |
97% | |
Air Sensitive | |
Barium hydroxide octahydrate |
Safety and Handling
GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H332-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 241-234-5
missing translation for 'rtecsNumber' : CQ9200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Barium hydroxide octahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : barium hydroxide octahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.