Learn More
Barium hydroxide octahydrate, ACS, 98+%, Thermo Scientific Chemicals
£29.75 - £80.90
Chemical Identifiers
CAS | 12230-71-6 |
---|---|
Molecular Formula | BaH18O10 |
Molecular Weight (g/mol) | 315.46 |
MDL Number | MFCD00149152 |
InChI Key | ZUDYPQRUOYEARG-UHFFFAOYSA-L |
Synonym | barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate |
PubChem CID | 17749109 |
SMILES | O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396479
|
Thermo Scientific Alfa Aesar
014499.30 |
250 g |
£29.75
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386479
|
Thermo Scientific Alfa Aesar
014499.A1 |
1 kg |
£80.90
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in manufacture of alkali, glass; in synthetic rubber vulcanization, alkalizing agent in water softening, boiler scale remedy, softening water.Barium hydroxide octahydrate is used in the centromeric heterochromatin banding technique. It acts as a precursor to prepare barium titanate, barium sulfate, barium phosphate and barium sulfide. Further, it is used in analytical chemistry for the titration of weak acids especially organic acids. It is involved in the preparation of cyclopentanone, diacetone alcohol and D-Gulonic gamma-lactone. In addition to this, it finds application in ester hydrolysis such as dimethyl hendecanedioate and decarboxylation of amino acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12230-71-6 | |
315.46 | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
17749109 |
BaH18O10 | |
MFCD00149152 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
Specifications
78°C | |
Crystalline | |
BaH18O10 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
barium(2+) octahydrate dihydroxide | |
17749109 | |
ACS Reagent | |
2.18 |
12230-71-6 | |
>98.0% | |
MFCD00149152 | |
Soluble in water and methanol. Slightly soluble in ethanol. Insoluble in acetone. | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] | |
315.46 | |
315.48 (171.36 Anhydrous) | |
Air sensitive; Hygroscopic | |
Barium hydroxide octahydrate, For ACS analysis |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H332-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 241-234-5
missing translation for 'rtecsNumber' : CQ9200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Barium hydroxide octahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Barium hydroxide octahydrate
RUO – Research Use Only